Merck
  • Supplemental dietary leucine and the skeletal muscle anabolic response to essential amino acids.
Supplemental dietary leucine and the skeletal muscle anabolic response to essential amino acids.
Nutrition reviews (2011-09-03)
Stefan M Pasiakos, James P McClung
ZUSAMMENFASSUNG
Skeletal muscle protein synthesis (MPS) is regulated by a number of dietary factors, to include essential amino acids (EAAs). Leucine, a branched-chain amino acid, has been identified as a stimulator of MPS in many cell culture and animal studies. However, whether supplemental leucine exerts a unique stimulatory effect, as compared to other EAAs, on muscle anabolism in humans has not been clearly demonstrated. A recent study found no improvement in resting MPS in adults who consumed a 10 g EAA supplement providing added leucine (3.5 g leucine) when compared to a control 10 g EAA supplement (1.8 g leucine). These findings suggest that added leucine is unnecessary for the stimulation of MPS when sufficient EAAs are provided; however, the study of supplemental leucine during conditions such as endurance exercise, caloric deprivation, and ageing may be warranted.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Leucin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Leucin, BioUltra, ≥99.5% (NT)
Supelco
L-Leucin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Leucin, 99%, FG
Leucin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Leucin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland