Merck
  • Reviewing the use of ethylcellulose, methylcellulose and hypromellose in microencapsulation. Part 3: Applications for microcapsules.
Reviewing the use of ethylcellulose, methylcellulose and hypromellose in microencapsulation. Part 3: Applications for microcapsules.
Drug development and industrial pharmacy (2011-10-04)
True L Rogers, Dave Wallick
ZUSAMMENFASSUNG
This three-part review has been developed following the evaluation of literature where ethylcellulose, methylcellulose, or hypromellose was used to make microcapsules. Parts 1 and 2 of the review are published in separate papers. Part 1 covers the various materials used to formulate microcapsules, and Part 2 covers the various techniques used to make microcapsules. In the current paper, Part 3 covers the end-use applications for which microcapsules are used. Examples of applications to be covered include modified release, improved efficacy and safety, multiparticulate compression, improved processability and stability, and taste- and odor-masking. It is hoped that formulators can use Part 3 to understand the various end-use applications of microcapsules made from these encapsulating polymers. SciFinder was utilized to perform the literature search. SciFinder leverages literature databases, such as Chemical Abstracts Service Registry and Medline. A total of 379 references were identified during the review. The need for a three-part review reflects the extensive amount of literature identified concerning these three encapsulating polymers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methylcellulose
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~120,000
USP
Hypromellose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~90,000