Merck
Bioconversion of crude glycerol by fungi.
Applied microbiology and biotechnology (2012-02-11)
R W Nicol, K Marchand, W D Lubitz
ZUSAMMENFASSUNG
The production of synthetic glycerol from petrochemical feedstocks has been decreasing in recent years. This is largely due to increasing supplies of crude glycerol derived as a co-product from the oleochemical industry, especially biodiesel production. The price of glycerol is at historic lows, and the supply of crude glycerol is projected to grow faster than its industrial uses. This oversupply is driving the transition from glycerol as a product to glycerol as a precursor for new industrial applications, including its use as a substrate for bioconversion. This article reviews the use of fungi for the bioconversion of crude glycerol to the value-added products 1,2-propanediol, ethanol, single cell oil, specialty polyunsaturated fatty acids, biosurfactants, and organic acids. Information on the impurities of crude glycerol from different industrial processes is also included.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Supelco
Glycerin, analytical standard
Sigma-Aldrich
Glycerin, tested according to Ph. Eur., anhydrous