Merck
[The microbiological transformation and degradation of pyridine and alkyl pyridines].
Nauchnye doklady vysshei shkoly. Biologicheskie nauki (1990-01-01)
E V Dovgilevich
ZUSAMMENFASSUNG
The scientific literary data on the microbiological transformation and degradation of pyridine, methylpyridines (picolines), dimethylpyridines (lutidines) and some ethylpyridines are summarized and analysed in this review. These highly toxic compounds are on the one hand the raw materials for medicinal preparation production with microorganisms, on the other hand they are industrial wastes polluting the environment which may be degraded microbiologically.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridin, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridin, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
Pyridin, ≥99%
Sigma-Aldrich
Pyridin, biotech. grade, ≥99.9%
Supelco
Pyridin, analytical standard
Supelco
Pyridin -Lösung, certified reference material, 2000 μg/mL in methanol