Merck
The Na⁺/L-proline transporter PutP.
Frontiers in bioscience (Landmark edition) (2011-12-29)
Heinrich Jung, Daniel Hilger, Michael Raba
ZUSAMMENFASSUNG
The Na⁺/L-proline transporter PutP is a member of the Na⁺/solute symporter family (TC 2A.21, SLC5), which contains several hundred proteins of pro- and eukaryotic origin. Within the family, the capability of L-proline uptake is restricted to proteins of prokaryotes. PutP contributes to the use of L-proline as a nutrient. In addition, the transporter may supply cells with compatible solute during adaptation to osmotic stress. Based on these and other functions, PutP is of significance for various bacteria-host interactions including the virulence of human pathogens. A homology model of Escherichia coli PutP was generated based on the crystal structure of the Vibrio parahaemolyticus Na+/galactose symporter. According to the model, PutP has a core structure of five plus five transmembrane domains forming an inverted repeat similar as originally revealed by the crystal structure of the Na+/leucine transporter LeuT. The homology model is experimentally verified by Cys cross-linking and site-directed spin labeling in combination with electron paramagnetic resonance spectroscopy. The putative sites of Na⁺ and L-proline binding are described, and a putative transport mechanism is discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Prolin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-Prolin, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Prolin
Sigma-Aldrich
L-Prolin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Prolin, 99%, FCC, FG
USP
L-Prolin, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Prolin, Pharmaceutical Secondary Standard; Certified Reference Material
Prolin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Prolin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland