Merck
  • Hypertonic saline, not mannitol, should be considered gold-standard medical therapy for intracranial hypertension.
Hypertonic saline, not mannitol, should be considered gold-standard medical therapy for intracranial hypertension.
Critical care (London, England) (2012-02-23)
Nicholas F Marko
ZUSAMMENFASSUNG
Hyperosmolar therapy is the principal medical management strategy for elevated intracranial pressure. Mannitol has been the primary hyperosmolar agent for nearly a century and remains the de facto gold standard for medical management of intracranial hypertension. Over the past 25 years, however, hypertonic saline (HTS) has become a progressively more common alternative to mannitol, and several recent studies have suggested its relative superiority. These findings have prompted calls for large-scale comparator trials of mannitol and HTS, but such trials would only be necessary if the designation of mannitol as the gold standard is appropriate and if current evidence suggests its therapeutic equipoise with HTS. Mounting evidence supporting HTS suggests that neither of these conditions is necessarily true and, instead, mandates reassessment of the actual gold-standard agent for hyperosmolar therapy. In the present article I make the case that current evidence supports HTS, not mannitol, as the better choice for gold-standard therapy for medical management of intracranial hypertension. This is accomplished first by examining the evidence on which the apparent designation of mannitol as the presumed gold-standard is based, then by reviewing the recent comparative efficacy data for HTS versus mannitol, and finally by discussing additional clinical considerations for appropriate designation of a gold-standard agent for hyperosmolar therapy. This assessment has important implications both for patient care and for clinical trial design.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Supelco
D-Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Mannitol, European Pharmacopoeia (EP) Reference Standard
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination