Merck
Extra-label use of ivermectin in some minor ruminant species: pharmacokinetic aspects.
Current pharmaceutical biotechnology (2011-11-02)
A González-Canga, R Belmar-Liberato, M Escribano
ZUSAMMENFASSUNG
The characterisation of ivermectin pharmacokinetics can be used to predict and to ensure an optimal activity in the target species and for designing programmes aimed for parasite control. Ivermectin pharmacokinetic studies performed in several minor ruminant species are reviewed in this paper with the aim of facilitating the adoption of rational basis for the establishment of appropriate dosage schedules.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ivermectin
Ivermectin, European Pharmacopoeia (EP) Reference Standard