Merck
In vitro testing of zinc oxide sunscreens.
International journal of cosmetic science (2012-05-18)
P G McCormick, T Tsuzuki
ZUSAMMENFASSUNG
The UVA performances of two all-mineral zinc oxide sunscreens are measured following Colipa and ISO procedures and compared to a sunscreen containing only organic actives. It is found that the two sunscreen types yield very different in vitro SPF and UVA results. It shown that the results can be rationalized in terms of the differences in the monochromatic extinction spectra of the two types of sunscreens.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zinkoxid, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Zinkoxid, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinkoxid, nanopowder, ≤50 nm particle size
Sigma-Aldrich
Zinkoxid, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Zinkoxid, Dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Zinkoxid, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Zinkoxid, 99.99% trace metals basis
Sigma-Aldrich
Zinkoxid, 99.999% trace metals basis
Sigma-Aldrich
Zinkoxid, nanowires, Length 1 um
Supelco
Zinkoxid, analytical standard