Merck
Regulatory roles for L-arginine in reducing white adipose tissue.
Frontiers in bioscience (Landmark edition) (2012-06-02)
Bi'e Tan, Xinguo Li, Yulong Yin, Zhenlong Wu, Chuang Liu, Carmen D Tekwe, Guoyao Wu
ZUSAMMENFASSUNG
As the nitrogenous precursor of nitric oxide, L-arginine regulates multiple metabolic pathways involved in the metabolism of fatty acids, glucose, amino acids, and proteins through cell signaling and gene expression. Specifically, arginine stimulates lipolysis and the expression of key genes responsible for activation of fatty acid oxidation to CO2 and water. The underlying mechanisms involve increases in the expression of peroxisome proliferator-activated receptor-gamma coactivator-1 alpha (PGC-1 alpha), mitochondrial biogenesis, and the growth of brown adipose tissue growth. Furthermore, arginine regulates adipocyte-muscle crosstalk and energy partitioning via the secretion of cytokines and hormones. In addition, arginine enhances AMP-activated protein kinase (AMPK) expression and activity, thereby modulating lipid metabolism and energy balance toward the loss of triacylglycerols. Growing evidence shows that dietary supplementation with arginine effectively reduces white adipose tissue in Zucker diabetic fatty rats, diet-induced obese rats, growing-finishing pigs, and obese patients with type II diabetes. Thus, arginine can be used to prevent and treat adiposity and the associated metabolic syndrome.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Arginin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginin, reagent grade, ≥98%
Sigma-Aldrich
L-Arginin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Arginin, 99%, FCC, FG
SAFC
L-Arginin
Supelco
L-Arginin, Pharmaceutical Secondary Standard; Certified Reference Material
Arginin, European Pharmacopoeia (EP) Reference Standard