Merck
Inhaled mannitol in the treatment of non-cystic fibrosis bronchiectasis in adults.
Respirology (Carlton, Vic.) (2012-05-09)
Juliana Gjoerup, Ole Hilberg, Elisabeth Bendstrup
ZUSAMMENFASSUNG
Bronchiectasis is a chronic disorder characterized by impaired mucociliary clearance and a relentless cycle of infection, inflammation and bronchial wall injury, which has a debilitating impact on the patient's quality of life and results in increased morbidity and mortality. It is a disease for which there are no currently registered therapeutic products, limited epidemiological data and no validated endpoints that have been accepted by the majority of regulatory authorities. This article reviews the evidence base for the efficacy of inhaled dry powder mannitol in patients with non-cystic fibrosis (CF) bronchiectasis. The few published papers on this topic concluded that mannitol, when inhaled as a dry powder, is a promising treatment for bronchiectasis. It is designed to hydrate the lungs and restore normal mucociliary clearance mechanisms in the lungs. Based on the available evidence, mannitol inhalation is well tolerated and improves the quality of life of patients with non-CF bronchiectasis. There is a need for well designed and adequately powered multicentre trials to establish the potential usefulness of mannitol as a treatment for non-CF bronchiectasis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Supelco
D-Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Mannitol, European Pharmacopoeia (EP) Reference Standard
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination