Merck
Aliskiren, amlodipine and hydrochlorothiazide triple combination for hypertension.
Expert review of cardiovascular therapy (2012-03-07)
Eric Judd, Edgar A Jaimes
ZUSAMMENFASSUNG
Cardiovascular-related morbidity and mortality is linked to hypertension with proportional gains in cardiovascular risk factor reduction with the lowering of blood pressure. Clinical trial data has shown that attaining goal blood pressure requires, for most patients, at least two antihypertensive medications, with a significant proportion requiring regimens of three or more medications. Single-pill triple combinations have returned to the market following results of increased efficacy and adherence over dual- and mono-therapy. The combination of aliskiren, amlodipine and hydrochlorothiazide is a rational choice for combination therapy and recent studies suggest that it is safe and effective in lowering blood pressure in patients who fail dual combination therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Hydrochlorthiazid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Hydrochlorthiazid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrochlorthiazid, crystalline
Hydrochlorthiazid, European Pharmacopoeia (EP) Reference Standard
Supelco
Hydrochlorothiazide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Hydrochlorthiazid, meets USP testing specifications
Hydrochlorothiazid für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard