Merck
Flurbiprofen.
Profiles of drug substances, excipients, and related methodology (2012-04-04)
Alaa A-M Abdel-Aziz, Abdullah A Al-Badr, Gamal Abdel Hafez
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Flurbiprofen, cyclooxygenase inhibitor
Flurbiprofen, European Pharmacopoeia (EP) Reference Standard