Merck
Genetic toxicology of thallium: a review.
Drug and chemical toxicology (2012-09-14)
Juan José Rodríguez-Mercado, Mario Agustín Altamirano-Lozano
ZUSAMMENFASSUNG
This review summarizes the current knowledge about the general toxicity of thallium (Tl) and its environmental sources, with special emphasis placed on its potential mutagenic, genotoxic, and cytotoxic effects on both eukaryotic and prokaryotic cells. Tl is a nonessential heavy metal that poses environmental and occupational threats as well as therapeutic hazards because of its use in medicine. It is found in two oxidation states, thallous (Tl(+)) and thallic (Tl(3+)), both of which are considered highly toxic to human beings and domestic and wild organisms. Many Tl compounds are colorless, odorless and tasteless, and these characteristics, combined with the high toxicity of TI compounds, have led to their use as poisons. Because of its similarity to potassium ions (K(+)), plants and mammals readily absorb Tl(+) through the skin and digestive and respiratory systems. In mammals, it can cross the placental, hematoencephalic, and gonadal barriers. Inside cells, Tl can accumulate and interfere with the metabolism of potassium and other metal cations, mimicking or inhibiting their action. The effects of Tl on genetic material have not yet been thoroughly explored, and few existing studies have focused exclusively on Tl(+). Both in vivo and in vitro studies indicate that Tl compounds can have a weak mutagenic effect, but no definitive effect on the induction of primary DNA damage or chromosomal damage has been shown. These studies have demonstrated that Tl compounds are highly toxic and lead to changes in cell-cycle progression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thallium, granular, 99.9% trace metals basis