Merck
  • Preoperative chlorhexidine shower or bath for prevention of surgical site infection: a meta-analysis.
Preoperative chlorhexidine shower or bath for prevention of surgical site infection: a meta-analysis.
American journal of infection control (2012-06-23)
Maciej Piotr Chlebicki, Nasia Safdar, John Charles O'Horo, Dennis G Maki
ZUSAMMENFASSUNG
Chlorhexidine showering is frequently recommended as an important preoperative measure to prevent surgical site infection (SSI). However, the efficacy of this approach is uncertain. A search of electronic databases was undertaken to identify prospective controlled trials evaluating whole-body preoperative bathing with chlorhexidine versus placebo or no bath for prevention of SSI. Summary risk ratios were calculated using a DerSimonian-Laird random effects model and a Mantel-Haenzel dichotomous effects model. Sixteen trials met inclusion criteria with a total of 17,932 patients: 7,952 patients received a chlorhexidine bath, and 9,980 patients were allocated to various comparator groups. Overall, 6.8% of patients developed SSI in the chlorhexidine group compared with 7.2% of patients in the comparator groups. Chlorhexidine bathing did not significantly reduce overall incidence of SSI when compared with soap, placebo, or no shower or bath (relative risk, 0.90; 95% confidence interval: 0.77-1.05, P = .19). Meta-analysis of available clinical trials suggests no appreciable benefit of preoperative whole-body chlorhexidine bathing for prevention of SSI. However, most studies omitted details of chlorhexidine application. Better designed trials with a specified duration and frequency of exposure to chlorhexidine are needed to determine whether preoperative whole-body chlorhexidine bathing reduces SSI.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chlorhexidin, ≥99.5%
Supelco
Chlorhexidin, Pharmaceutical Secondary Standard; Certified Reference Material
Chlorhexidin, European Pharmacopoeia (EP) Reference Standard