Merck
Oxaliplatin-induced thrombotic thrombocytopenic purpura: case report and literature review.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2012-09-19)
Jiaxin Niu, Martha P Mims
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oxaliplatin, powder
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard