Merck
  • [Advances in the research of rapid detection methods of pyrazinamide-resistant Mycobacterium tuberculosis].
[Advances in the research of rapid detection methods of pyrazinamide-resistant Mycobacterium tuberculosis].
Zhonghua jie he he hu xi za zhi = Zhonghua jiehe he huxi zazhi = Chinese journal of tuberculosis and respiratory diseases (2012-07-12)
Chuang-yue Hong, Xiao-li Liu, Feng Wang
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyrazincarboxamid
Pyrazinamid, European Pharmacopoeia (EP) Reference Standard