Merck
Role of glutathione in winemaking: a review.
Journal of agricultural and food chemistry (2012-12-18)
Engela C Kritzinger, Florian F Bauer, Wessel J du Toit
ZUSAMMENFASSUNG
Glutathione is an important constituent of grapes, must, and wine. However, to date, no review has provided an integrated view of the role of this compound in wine-related systems. In this review, special emphasis is given to its occurrence in grapes, must, and wine and its role as an antioxidant in wine. The effect of glutathione on both desirable and undesirable aroma compounds is also outlined. Furthermore, the use of glutathione-enriched products in winemaking and the various analytical techniques for the quantification of glutathione in must and wine are discussed. Limitations in existing knowledge are also identified.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Supelco
Glutathion, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Glutathion, European Pharmacopoeia (EP) Reference Standard