Merck
Strontium biokinetic model for mouse-like rodent.
Journal of environmental radioactivity (2012-12-19)
Georgy Malinovsky, Ilia Yarmoshenko, Michael Zhukovsky, Vera Starichenko, Makar Modorov
ZUSAMMENFASSUNG
Model describing the biokinetics of strontium for murine rodent is suggested. The model represents modification of the ICRP model for reference human with reduced number of compartments: Blood, Gastrointestinal tract, Soft tissues, Skeleton, Urinary bladder. To estimate transfer rates of the model the published experimental data on strontium retention in body of laboratory and wild mice were analyzed. A set of eleven transfer rates suggested for the strontium biokinetic model for murine rodent satisfactorily describes both the laboratory experiments (relative standard error of 9.5%) and data on radiostrontium content available for wild animals. Application of the model allows estimation of strontium distribution by organs and tissues both in the cases of acute and chronic exposure with assessment of strontium activity in organs with time since beginning of exposure. The developed strontium biokinetic model will be used for internal dose assessment for murine rodents inhabiting East-Ural Radioactive Trace, where (90)Sr intake is a significant source of contemporary internal exposure.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Strontium, granular, 99% trace metals basis
Sigma-Aldrich
Strontium, random pieces, 99%
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.9% trace metals basis