Merck
Stability, transdermal penetration, and cutaneous effects of ascorbic acid and its derivatives.
Journal of cosmetic dermatology (2012-11-24)
Nicholas P J Stamford
ZUSAMMENFASSUNG
Topically applied antioxidants exert their benefits by offering protection from damaging free radicals and over-the-counter cosmeceuticals incorporating antioxidants are among the most popular anti-aging products available. One potent antioxidant of particular note, vitamin C, has been extensively utilized because it possesses a variety of other cutaneous benefits including photoprotection from UV A & B, neocollagenesis, inhibition of melanogenesis and improvement of a variety of inflammatory skin disorders. However, the instability of this water-soluble vitamin, together with difficulties associated with its topical delivery, has presented issues for the formulation chemist. This article reviews the scientific data and clinical studies that underpin the stability, percutaneous absorption, and cutaneous effects of vitamin C together with its commonly utilized, commercially available derivatives.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ascorbinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG
Sigma-Aldrich
L-Ascorbinsäure, BioUltra, ≥99.5% (RT)
Ascorbinsäure, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbinsäure, tested according to Ph. Eur.