Merck
All-trans retinoic acid in the treatment of pediatric acute promyelocytic leukemia.
Expert review of anticancer therapy (2012-10-27)
Riccardo Masetti, Francesca Vendemini, Daniele Zama, Carlotta Biagi, Pietro Gasperini, Andrea Pession
ZUSAMMENFASSUNG
Acute promyelocytic leukemia (APL) is a rare form of acute myeloid leukemia with specific epidemiological, pathogenetic and clinical features. Its frequency varies widely among nations, with a decreased incidence among 'Nordic' origin populations. The molecular hallmark of the disease is the presence of a balanced reciprocal translocation resulting in the PML/RAR-α gene fusion, which represents the target of the all-trans retinoic acid (ATRA) therapy. The introduction of ATRA in conjunction with anthracyclines marked a turning point in the treatment of APL, previously associated with a significant morbidity and mortality. Nowadays the standard front-line therapy for pediatric APL includes ATRA in every phase of the treatment, resulting in a complete remission rate of 90-95%. Here we provide an overview of the role of ATRA in the treatment of pediatric APL, summarizing the most relevant clinical results of recent decades and investigating future therapeutic perspectives for children with APL.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Supelco
Retinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Tretinoin, United States Pharmacopeia (USP) Reference Standard
Tretinoin, European Pharmacopoeia (EP) Reference Standard