Merck
Molecular engineering of guanine-rich sequences: Z-DNA, DNA triplexes, and G-quadruplexes.
Chemical reviews (2013-02-09)
Osman Doluca, Jamie M Withers, Vyacheslav V Filichev
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Guanin, 98%
Supelco
Guanin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Guanin, BioUltra