Merck
  • Ethinyl estradiol and 17β-estradiol in combined oral contraceptives: pharmacokinetics, pharmacodynamics and risk assessment.
Ethinyl estradiol and 17β-estradiol in combined oral contraceptives: pharmacokinetics, pharmacodynamics and risk assessment.
Contraception (2013-02-05)
Frank Z Stanczyk, David F Archer, Bhagu R Bhavnani
ZUSAMMENFASSUNG
The need to seek improved combined oral contraceptive (COC) efficacy, with fewer health risks and better acceptability, has been ongoing since the introduction of COCs more than 50 years ago. New progestin formulations combined with lower doses of ethinyl estradiol (EE), the predominant estrogenic component of COCs, have reduced the incidence of venous thromboembolism and other negative outcomes of COC treatment. Previous attempts to use endogenous 17β-estradiol (E₂) instead of EE were limited primarily by poor cycle control. The recent introduction of E₂-based formulations has renewed interest to determine if there are potential benefits of using E₂ in COCs. These formulations have been shown to have similar efficacy and cycle control as EE-based COCs. This review provides a brief summary of the pharmacology of EE and E₂, including metabolism, pharmacokinetics and pharmacodynamics, as well as adverse effects of these estrogens.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
β-Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
17α-Ethinylestradiol, ≥98%
Supelco
17β-Estradiol -Lösung, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ethinylestradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
β-Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Estradiol, analytical standard
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
USP
Ethinylestradiol, United States Pharmacopeia (USP) Reference Standard
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
Supelco
17α-Ethynylestradiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Ethinylestradiol für die Systemeignung, European Pharmacopoeia (EP) Reference Standard