Merck
  • Association of arsenic, cadmium and manganese exposure with neurodevelopment and behavioural disorders in children: a systematic review and meta-analysis.
Association of arsenic, cadmium and manganese exposure with neurodevelopment and behavioural disorders in children: a systematic review and meta-analysis.
The Science of the total environment (2013-04-11)
Miguel Rodríguez-Barranco, Marina Lacasaña, Clemente Aguilar-Garduño, Juan Alguacil, Fernando Gil, Beatriz González-Alzaga, Antonio Rojas-García
ZUSAMMENFASSUNG
The aim of this study was to analyse the scientific evidence published to date on the potential effects on neurodevelopment and behavioural disorders in children exposed to arsenic, cadmium and manganese and to quantify the magnitude of the effect on neurodevelopment by pooling the results of the different studies. We conducted a systematic review of original articles from January 2000 until March 2012, that evaluate the effects on neurodevelopment and behavioural disorders due to pre or post natal exposure to arsenic, cadmium and manganese in children up to 16 years of age. We also conducted a meta-analysis assessing the effects of exposure to arsenic and manganese on neurodevelopment. Forty-one articles that evaluated the effects of metallic elements on neurodevelopment and behavioural disorders met the inclusion criteria: 18 examined arsenic, 6 cadmium and 17 manganese. Most studies evaluating exposure to arsenic (13 of 18) and manganese (14 of 17) reported a significant negative effect on neurodevelopment and behavioural disorders. Only two studies that evaluated exposure to cadmium found an association with neurodevelopmental or behavioural disorders. The results of our meta-analysis suggest that a 50% increase of arsenic levels in urine would be associated with a 0.4 decrease in the intelligence quotient (IQ) of children aged 5-15 years. Moreover a 50% increase of manganese levels in hair would be associated with a decrease of 0.7 points in the IQ of children aged 6-13 years. There is evidence that relates arsenic and manganese exposure with neurodevelopmental problems in children, but there is little information on cadmium exposure. Few studies have evaluated behavioural disorders due to exposure to these compounds, and manganese is the only one for which there is more evidence of the existence of association with attention deficit disorder with hyperactivity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mangan, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Arsen, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Arsen, powder, ≥99.997% trace metals basis
Sigma-Aldrich
Mangan, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Mangan, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 100%
Mangan, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Mangan, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Cadmium, rod, 100mm, diameter 40mm, 99.9%
Cadmium, rod, 25mm, diameter 40mm, 99.9%
Mangan, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Mangan, rod, 10mm, diameter 4.0mm, cast, 99.5%
Mangan, rod, 50mm, diameter 4.0mm, cast, 99.5%
Cadmium, foil, 2m coil, thickness 0.5mm, as rolled, 100%
Cadmium, wire reel, 1m, diameter 1.2mm, as drawn, 99.99+%
Mangan, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 98.7%
Cadmium, rod, 200mm, diameter 10.0mm, 99.999%
Cadmium, rod, 50mm, diameter 10.0mm, 99.999%
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Mangan, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%