Merck
Methionine oxidation and reduction in proteins.
Biochimica et biophysica acta (2013-05-08)
Geumsoo Kim, Stephen J Weiss, Rodney L Levine
ZUSAMMENFASSUNG
Cysteine and methionine are the two sulfur containing amino acids in proteins. While the roles of protein-bound cysteinyl residues as endogenous antioxidants are well appreciated, those of methionine remain largely unexplored. We summarize the key roles of methionine residues in proteins. Recent studies establish that cysteine and methionine have remarkably similar functions. Both cysteine and methionine serve as important cellular antioxidants, stabilize the structure of proteins, and can act as regulatory switches through reversible oxidation and reduction. This article is part of a Special Issue entitled Current methods to study reactive oxygen species - pros and cons and biophysics of membrane proteins. Guest Editor: Christine Winterbourn.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Methionin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SAFC
L-Methionin
Sigma-Aldrich
L-Methionin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionin, reagent grade, ≥98% (HPLC)
Supelco
L-Methionin, Pharmaceutical Secondary Standard; Certified Reference Material
Methionin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Methionin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland