Merck
Properties, metabolisms, and applications of (L)-proline analogues.
Applied microbiology and biotechnology (2013-06-20)
Thi Mai Hoa Bach, Hiroshi Takagi
ZUSAMMENFASSUNG
Due to the unique role of L-proline in the folding and structure of protein, a variety of synthetic proline analogues have been developed. L-Proline analogues have been proven to be valuable reagents for studying cellular metabolism and the regulation of macromolecule synthesis in both prokaryotic and eukaryotic cells. In addition to these fundamental researches, they are useful compounds for industrial use. For instance, microorganisms that overproduce L-proline have been obtained by isolating mutants resistant to L-proline analogues. They are also promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occurring or de novo designed peptides. Among L-proline analogues, L-azetidine-2-carboxylic acid (L-AZC) is a toxic non-proteinogenic amino acid originally found in lily of the valley plants and trans-4-hydroxy-L-proline (4-L-THOP) is the most abundant component of mammalian collagen. Many hydroxyprolines (HOPs), such as 4-L-THOP and cis-4-hydroxy-L-proline (4-L-CHOP), are useful chiral building blocks for the organic synthesis of pharmaceuticals. In addition, L-AZC and 4-L-CHOP, which are potent inhibitors of cell growth, have been tested for their antitumor activity in tissue culture and in vivo. In this review, we describe the recent discoveries regarding the physiological properties and microbial production and metabolism of L-proline analogues, particularly L-AZC and HOPs. Their applications in fundamental research and industrial use are also discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Prolin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-Prolin, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Prolin
Sigma-Aldrich
L-Prolin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Prolin, 99%, FCC, FG
USP
L-Prolin, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Prolin, Pharmaceutical Secondary Standard; Certified Reference Material
Prolin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Prolin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland