Merck
Acetazolamide-responsive ataxia.
Seminars in neurology (2013-05-17)
Vikas Kotagal
ZUSAMMENFASSUNG
Acetazolamide-responsive ataxia represents a unique collection of genetically distinct episodic ataxia (EA) disorders associated with paroxysmal cerebellar symptoms many of which are responsive to medical treatment with acetazolamide, a carbonic anhydrase inhibitor. Among all of the subtypes of episodic ataxia, types 2 (EA2), 3 (EA3), and 5 (EA5) are thought be the most medication responsive. Some patients with episodic ataxia type 1 (EA1) will also describe improvement with acetazolamide. Each of these individual genetic syndromes is characterized by its own unique mechanism and clinical presentation. In this review, the author provides an overview of the pathophysiology of acetazolamide-responsive ataxia, its natural history, and its clinical management.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetazolamid, ≥99%, powder
USP
Acetazolamid, United States Pharmacopeia (USP) Reference Standard
Acetazolamid, European Pharmacopoeia (EP) Reference Standard
Acetazolamid für die Systemeignung, European Pharmacopoeia (EP) Reference Standard