Merck
[Ursolic acid as antitumor agent and inductor of PTEN and brown fat].
Voprosy onkologii (2012-01-01)
L M Bershteĭn
ZUSAMMENFASSUNG
In this mini-review the basic evidence about anticancer properties of ursolic acid (UA), the compound belonging to the class of triterpenoids, is given. Beside inhibiting tumor cell growth in vitro and in vivo and activating of apoptosis, UA (as well as some other related and not related compounds) is capable to induce PTEN (a tumor suppressor mutation of which is rather often discovered in human tumors including endometrial cancer type I) and amount/activity of brown fat. The latter action may explain obesity-preventing capacity of UA that also may lead to an additional antiblastomogenic effect.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ursolsäure, ≥90%
Supelco
Ursolsäure, analytical standard
Ursolsäure, primary reference standard
Ursolsäure, European Pharmacopoeia (EP) Reference Standard