Merck
Retinoic acid signaling in spinal cord development.
The international journal of biochemistry & cell biology (2013-04-13)
Ricardo Lara-Ramírez, Elisabeth Zieger, Michael Schubert
ZUSAMMENFASSUNG
Retinoic acid (RA) is an important signaling molecule mediating intercellular communication through vertebrate development. Here, we present and discuss recent information on the roles of the RA signaling pathway in spinal cord development. RA is an important player in the patterning and definition of the spinal cord territory from very early stages of development, even before the appearance of the neural plate and further serves a role in the patterning of the spinal cord both along the dorsoventral and anteroposterior axes, particularly in the promotion of neuronal differentiation. It is thus required to establish a variety of neuronal cell types at specific positions of the spinal cord. The main goal of this review is to gather information from vertebrate models, including fish, frogs, chicken and mice, and to put this information in a comparative context in an effort to visualize how the RA pathway was incorporated into the evolving vertebrate spinal cord and to identify mechanisms that are both common and different in the various vertebrate models. In doing so, we try to reconstruct how spinal cord development has been regulated by the RA signaling cascade through vertebrate diversification, highlighting areas which require further studies to obtain a better understanding of the evolutionary events that shaped this structure in the vertebrate lineage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Supelco
Retinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Tretinoin, United States Pharmacopeia (USP) Reference Standard
Tretinoin, European Pharmacopoeia (EP) Reference Standard