Merck
[Propofol up2date].
Anasthesiologie, Intensivmedizin, Notfallmedizin, Schmerztherapie : AINS (2013-08-10)
Stefan Bushuven, Daniel Heise
ZUSAMMENFASSUNG
Propofol is a common hypnotic agent in anaesthesiology and intensive care medicine and for procedural sedation as well. Despite the intensive usage the full potential, pharmacokinetic and -dynamic abilities, interactions and side effects of the substance may not be realized by all providers. Additionally there is dispute and unwarranted myth about the substance. Propofol is a highly potent hypnotic with a wide range of (un)desired effects. This article is reviewing the diversity of the substance in clinical practice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Propofol, European Pharmacopoeia (EP) Reference Standard