Merck
Cardiovascular effects of arsenic: clinical and epidemiological findings.
Environmental science and pollution research international (2013-09-11)
Francesco Stea, Fabrizio Bianchi, Liliana Cori, Rosa Sicari
ZUSAMMENFASSUNG
Several population studies relate exposure to high levels of arsenic with an increased incidence of ischemic heart disease and cardiovascular mortality. An association has been shown between exposure to high levels of arsenic and cardiovascular risk factors such as hypertension and diabetes mellitus, and vascular damage such as subclinical carotid atherosclerosis. The mechanisms underlying these phenomena are currently being studied and appear to indicate an alteration of vascular function. However, the effects of low levels of exposure to arsenic and their potential detrimental cardiovascular effect are less explored. The article provides an overview of the pathophysiologic mechanisms linking low-level arsenic exposure to the occurrence of cardiovascular disease and its complications, and some potential preventive strategies to implement.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Arsen, 99.999% trace metals basis
Sigma-Aldrich
Arsen, powder, ≥99.997% trace metals basis