Merck
Serine, glycine and one-carbon units: cancer metabolism in full circle.
Nature reviews. Cancer (2013-07-05)
Jason W Locasale
ZUSAMMENFASSUNG
One-carbon metabolism involving the folate and methionine cycles integrates nutritional status from amino acids, glucose and vitamins, and generates diverse outputs, such as the biosynthesis of lipids, nucleotides and proteins, the maintenance of redox status and the substrates for methylation reactions. Long considered a 'housekeeping' process, this pathway has recently been shown to have additional complexity. Genetic and functional evidence suggests that hyperactivation of this pathway is a driver of oncogenesis and establishes a link to cellular epigenetic status. Given the wealth of clinically available agents that target one-carbon metabolism, these new findings could present opportunities for translation into precision cancer medicine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L-Serin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
Glycin
Supelco
Glycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycin, 99%, FCC
Sigma-Aldrich
L-Serin, BioUltra, ≥99.5% (NT)
USP
Glycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycin 1 M -Lösung
Supelco
L-Serin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycin, analytical standard, for nitrogen determination according to Kjeldahl method
Glycin, European Pharmacopoeia (EP) Reference Standard
Serin, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycin, tested according to Ph. Eur.
Supelco
L-Serin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland