Merck
  • Nicotinamide riboside, a trace nutrient in foods, is a vitamin B3 with effects on energy metabolism and neuroprotection.
Nicotinamide riboside, a trace nutrient in foods, is a vitamin B3 with effects on energy metabolism and neuroprotection.
Current opinion in clinical nutrition and metabolic care (2013-09-28)
Yuling Chi, Anthony A Sauve
ZUSAMMENFASSUNG
This review focuses upon the biology and metabolism of a trace component in foods called nicotinamide riboside. Nicotinamide riboside is a precursor of nicotinamide adenine dinucleotide (NAD), and is a source of Vitamin B3. Evidence indicates that nicotinamide riboside has unique properties as a Vitamin B3. We review knowledge of the metabolism of this substance, as well as recent work suggesting novel health benefits that might be associated with nicotinamide riboside taken in larger quantities than is found naturally in foods. Recent work investigating the effects of nicotinamide riboside in yeast and mammals established that it is metabolized by at least two types of metabolic pathways. The first of these is degradative and produces nicotinamide. The second pathway involves kinases called nicotinamide riboside kinases (Nrk1 and Nrk2, in humans). The likely involvement of the kinase pathway is implicated in the unique effects of nicotinamide riboside in raising tissue NAD concentrations in rodents and for potent effects in eliciting insulin sensitivity, mitochondrial biogenesis, and enhancement of sirtuin functions. Additional studies with nicotinamide riboside in models of Alzheimer's disease indicate bioavailability to brain and protective effects, likely by stimulation of brain NAD synthesis. Initial studies have clarified the potential for a lesser-known Vitamin B3 called nicotinamide riboside that is available in selected foods, and possibly available to humans by supplements. It has properties that are insulin sensitizing, enhancing to exercise, resisting to negative effects of high-fat diet, and neuroprotecting.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nicotinamid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamid, ≥99.5% (HPLC)
Supelco
Niacinamid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nicotinamid, ≥98% (HPLC), powder
USP
Niacinamid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Niacinamid, meets USP testing specifications
Supelco
Nicotinamid (Vitamin B3) -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Nicotinamid, ≥98.5% (HPLC)
Nicotinamid, European Pharmacopoeia (EP) Reference Standard