Merck
Translational research investigations on ATP7A: an important human copper ATPase.
Annals of the New York Academy of Sciences (2014-04-17)
Stephen G Kaler
ZUSAMMENFASSUNG
In more than 40 years since copper deficiency was delineated in pediatric subjects with Menkes disease, remarkable advances in our understanding of the clinical, biochemical, and molecular aspects of the human copper transporter ATP7A have emerged. Mutations in the gene encoding this multitasking molecule are now implicated in at least two other distinctive phenotypes: occipital horn syndrome and ATP7A-related isolated distal motor neuropathy. Several other novel inherited disorders of copper metabolism have been identified in the past several years, aided by advances in human gene mapping and automated DNA sequencing. In this paper, I review the history and evolution of our understanding of disorders caused by impaired ATP7A function, and outline future challenges.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kupfer, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Kupfer, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Kupfer, powder, 99.99% trace metals basis
Sigma-Aldrich
Kupfer, powder, <75 μm, 99%
Sigma-Aldrich
Kupfer, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Kupfer, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Kupfer, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
Kupfer, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Kupfer, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Kupfer, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Kupfer, nanopowder, 60-80 nm particle size (APS), ≥99.5% trace metals basis
Sigma-Aldrich
Kupfer, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Kupfer, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Kupfer, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Kupfer, foil, thickness 1.0 mm, 99.999% trace metals basis
Kupfer, mesh, 100x100mm, nominal aperture 0.14mm, thickness 0.25mm, wire diameter 0.115mm, 100x100 wires/inch, open area 30.3%, plain weave mesh
Sigma-Aldrich
Kupfer, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Kupfer, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Kupfer, shot, −3-+14 mesh, 99%
Sigma-Aldrich
Kupfer, beads, 2-8 mm, ≥99.99% trace metals basis
Kupfer, wire reel, 100m, diameter 0.05mm, annealed, 99.9%
Sigma-Aldrich
Kupfer, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Kupfer, wire, diam. 0.64 mm, 99.995% trace metals basis
Kupfer, sheet, 100x100mm, thickness 6.35mm, 99.95+%
Kupfer, rod, 500mm, diameter 2.0mm, as drawn, 99.99+%
Kupfer, rod, 1000mm, diameter 4.8mm, hard, 99.9%
Kupfer, rod, 100mm, diameter 9.5mm, hard, 99.9%
Kupfer, rod, 50mm, diameter 6.35mm, as drawn, 99.999%
Kupfer, mesh, 600x600mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Kupfer, foil, 100x100mm, thickness 0.1mm, hard, 99.95+%