Merck
  • Carbon dioxide laser versus stapler-assisted endoscopic Zenker's diverticulotomy: a systematic review and meta-analysis.
Carbon dioxide laser versus stapler-assisted endoscopic Zenker's diverticulotomy: a systematic review and meta-analysis.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery (2014-02-06)
Noah P Parker, Stephanie Misono
ZUSAMMENFASSUNG
To evaluate outcomes following endoscopic management of Zenker's diverticula using a carbon dioxide laser (CO2) or stapler-assisted technique, a systematic review and meta-analysis were conducted. Seven retrospective, uncontrolled case series including 391 procedures met selection criteria. No higher quality studies were identified. Outcomes favoring the stapler technique included a shorter duration of nil per os (NPO) status (2 studies), length of hospitalization (LOH, 2 studies), and fewer postoperative fevers and abnormal chest x-rays (1 study). Outcomes favoring the CO2 technique included greater improvement in postoperative dysphagia and regurgitation scores (2 studies) and a lower revision rate (1 study). Meta-analysis demonstrated increased nondental complications in the CO2 group (odds ratio 3.81; 95% confidence interval, 1.37-10.59; P = .01) but no difference in duration of NPO (P = .06), LOH (P = .07), overall complications (P = .08), dental complications (P = .57), major complications (P = .38), or revision surgery (P = .82). Implications are limited by the quality of studies identified.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kohlenstoffdioxid, ≥99.8%
Sigma-Aldrich
Kohlenstoff-12C-dioxid, 99.9 atom % 12C
Sigma-Aldrich
Kohlenstoff-12C-dioxid, 99.99 atom % 12C