Merck
[Morphine antagonists (pentazocine, nalorphine, naloxone and buprenophine)].
Cahiers d'anesthesiologie (1988-05-01)
T Fournier, P Besserve, P Lefèvre, A Lephay, M Ossart
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Pentazocine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®