Merck
Enzymes in the D-alanine branch of bacterial cell wall peptidoglycan assembly.
The Journal of biological chemistry (1989-02-15)
C T Walsh
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Alanin, ≥98% (TLC)
Sigma-Aldrich
L-Alanin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Alanin, ≥99%
Supelco
L-Alanin, Pharmaceutical Secondary Standard; Certified Reference Material
Alanin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Alanin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Alanin-12C3, 99.9 atom % 12C