Merck
Reappraisal of the risk/benefit of nitrofurantoin: review of toxicity and efficacy.
Adverse drug reactions and acute poisoning reviews (1989-01-01)
R R Shah, G Wade
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV), meets EP testing specifications
USP
Nitrofurantoin, United States Pharmacopeia (USP) Reference Standard
Supelco
Nitrofurantoin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Nitrofurantoin, VETRANAL®, analytical standard