Merck
  • Clinical significance of differences in bioavailability of medroxyprogesterone acetate preparations.
Clinical significance of differences in bioavailability of medroxyprogesterone acetate preparations.
Clinical pharmacokinetics (1989-03-01)
A D Stockdale, A Y Rostom
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Medroxyprogesteron