Merck
[Report on an autopsy of a cancer patient with Carmofur (HCFU) leukoencephalopathy].
Rinsho shinkeigaku = Clinical neurology (1987-11-01)
A Furuya, H Shiroyama, Y Yano, S Kishida, R Okeda
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Carmofur, ≥98% (HPLC), powder