Merck
  • Toxicological quandary of the use of bis (2-diethylhexyl) phthalate (DEHP) as a plasticizer for blood bags.
Toxicological quandary of the use of bis (2-diethylhexyl) phthalate (DEHP) as a plasticizer for blood bags.
Annals of clinical and laboratory science (1988-03-01)
B A Myhre
ZUSAMMENFASSUNG
Plastic bags are very useful containers for the storage of blood and blood products since they are relatively transparent, hard to break, can be sealed aseptically with a radio-frequency current, and can be centrifuged for the isolation of blood components. In order to make the plastic more flexible, various agents are added, of which the most common is di- (2-ethylhexyl) phthalate (DEHP). This plasticizer has been found to leach from the plastic into the blood components during the storage period. Some animal studies have shown that this chemical can produce cancers and various tissue abnormalities. The human data from multi-transfused patients do not clearly indicate any specific damage: however, because of the animal studies, work has been carried out to find a non-leachable plasticizer. Several have been found: unfortunately, when survival studies are done, the red cell life span of the stored blood is decreased. Current work seems to indicate that DEHP has a membrane stabilizing function that prolongs the storage time of the red cell. Therefore, there currently is a trade off between plasticizer presence and red cell life span that must be considered when designing new blood storage bags.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phthalsäure-bis-2-ethylhexylester, ≥99.5%
Supelco
Phthalsäure-bis-2-ethylhexylester, PESTANAL®, analytical standard
Supelco
Di-(2-ethylhexyl)-phthalat -Lösung, certified reference material, 2000 μg/mL in methanol
Supelco
Phthalsäure-bis-2-ethylhexylester, Selectophore
Supelco
Phthalsäure-bis-2-ethylhexylester, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Kunststoffzusatzstoff 01, European Pharmacopoeia (EP) Reference Standard