- [Metabolism of antipyrine in man].
[Metabolism of antipyrine in man].
Pathologie-biologie (1985-09-01)
F Pautet, R Barret, M Daudon, B Mathian
PMID3909077
ZUSAMMENFASSUNG
Antipyrine is widely used as a model compound to assess the hepatic metabolism in man. This drug is extensively metabolized by the hepatic microsomal drug-oxidizing system, into three main metabolites and six minor compounds of the phase I. The metabolite fate of this drug is not fully understood. A new metabolite pathway including a ring opening seems possible.