Merck
Indium-111 imaging in osteomyelitis and neuroarthropathy. Review and case report.
Journal of the American Podiatric Medical Association (1986-01-01)
L M Techner, C A Eiser, W Laine
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Indium, powder, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, pieces, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 0.45-0.55 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, bars, 99.95%
Sigma-Aldrich
Indium, wire, diam. 2.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis
Indium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.999%
Indium, foil, not light tested, 50x50mm, thickness 0.02mm, 99.8%
Indium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.999%
Indium, foil, 8mm disks, thickness 0.0125mm, temporary acrylic support, 99.8%
Indium, foil, 150x150mm, thickness 0.1mm, as rolled, 100%
Indium, wire reel, 2m, diameter 0.5mm, as drawn, 99.999%
Indium, wire reel, 2m, diameter 1.0mm, as drawn, 99.999%
Indium, foil, 100x200mm, thickness 0.5mm, as rolled, 99.999%
Indium, wire reel, 0.1m, diameter 1.8mm, as drawn, 99.999%
Indium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Indium, wire reel, 0.1m, diameter 1.5mm, as drawn, 99.999%
Indium, foil, not light tested, 25x25mm, thickness 0.05mm, as rolled, 100%
Indium, wire reel, 1m, diameter 1.0mm, as drawn, 99.999%
Indium, wire reel, 1m, diameter 1.5mm, as drawn, 99.999%