Merck
  • Induction and repair of benzo[a]pyrene-DNA adducts in C57BL/6 and BALB/c mice: association with aging and longevity.
Induction and repair of benzo[a]pyrene-DNA adducts in C57BL/6 and BALB/c mice: association with aging and longevity.
Mechanisms of ageing and development (1995-07-28)
M E Boerrigter, J Y Wei, J Vijg
ZUSAMMENFASSUNG
In this study, we employed the sensitive 32P-postlabeling assay to assess the influence of age on the formation and disappearance of benzo[a]pyrene (B[a]P) DNA adducts in six organs of two different mouse strains with different life spans, C57BL/6ByJ (C57BL/6) and BALB/cByJ (BALB/c). Following a single, intraperitoneal treatment with 50 mg B[a]P per kg of bodyweight, maximum formation of the major B[a]P-derived adduct, trans-(7R)-N2-[10-(7 beta,8 alpha, 9 alpha-trihydroxy- 7,8,9,10)-tetrahydrobenzo[a]pyrene]-yl-deoxyguanosine (BPDE-N2-dG), appeared to be age- and organ-dependent; minor differences were observed for the same organs between the two mouse strains. The maximum formation of BPDE-N2-dG in the various organs from young and old mice differed by a factor of 2-4 and was two- to eightfold lower in organs from old mice as compared to young mice. The removal of BPDE-N2-dG, up to 7 days after the treatment, was apparently age- and strain-dependent; non-significant differences were observed for organs within strains at each age studied. In young C57BL/6 mice, which have a greater life expectancy than BALB/c, the rate of disappearance of BPDE-N2-dG was significantly higher in liver and heart as compared to young BALB/c. At the older age a decrease in the rate of BPDE-N2-dG disappearance was observed more frequently, and to a relatively greater extent, in organs from C57BL/6 mice as compared to BALB/c mice. These results are discussed in relation to the differences in life spans and the incidence of pathological lesions between the two strains of mice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzo[a]pyren, ≥96% (HPLC)
Supelco
Benzo[a]pyren, analytical standard, for environmental analysis
Supelco
Benzo[a]pyren, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzo[a]pyren -Lösung, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
Benzo[a]pyren -Lösung, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Supelco
Benzo[a]pyren -Lösung, 100 μg/mL in cyclohexane, analytical standard