Merck
Neurotoxic reaction to lindane in an HIV-seropositive patient. An old medication's new problem.
The Journal of family practice (1995-03-01)
B A Solomon, S R Haut, E M Carr, A R Shalita
ZUSAMMENFASSUNG
Scabies is a common infestation for patients of all ages throughout the world. One of the standard therapies for scabies is 1% lindane lotion. Lindane has been associated with neurotoxic reactions, specifically seizures. We describe a case of a middle-aged adult man with human immunodeficiency virus (HIV) infection who was found to have typical scabies and was treated with a single topical application of lindane. Two hours after the application, the patient experienced a new-onset generalized seizure. We believe that the triad of HIV infection, medications that reduce seizure thresholds, and percutaneous absorption factors, in the aggregate, induced the seizure. We believe that lindane should not be prescribed for patients with HIV infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Lindan, PESTANAL®, analytical standard
Supelco
Lindan, Pharmaceutical Secondary Standard; Certified Reference Material