Merck
Kinetics and metabolism of pyrazolones (propyphenazone, aminopyrine and dipyrone).
British journal of clinical pharmacology (1980-10-01)
M Volz, H M Kellner
ZUSAMMENFASSUNG
1 Propyphenazone 220 mg was administered orally to volunteers. Maximum plasma concentrations between 1.5 microgram/ml and 3.5 micrograms/ml were found 30 min later. After comparable doses plasma concentrations in dog and rabbit were lower. The distribution volumes were 2 l/kg. 2 The major metabolic route of propyphenazone is demethylation. The main urinary metabolite is the enolglucuronide of N-(2)-demethylpropyphenazone. 3 Aminopyrine is rapidly and almost completely absorbed after oral administration. Maximum plasma concentrations of 10 microgram/ml are reached 1.5 h after a 500 mg dose. The biological half-life is 2-3 h, the relative distribution volume 60% on average, and binding to plasma proteins approximately 15%. 4 Unchanged aminopyrine is only excreted in small quantities. The major routes of metabolism are demethylation (4-methylaminoantipyrine and 4-aminoantipyrine) and acylation (4-acetyl and 4-formylaminoantipyrine). There are other biotransformation products. 5 After oral administration of [14C]-dipyrone 480 mg the maximum serum concentration of 13.4 +/- 0.8 microgram/ml occurred at 1-1.5 hours. 6 Dipyrone was not detectable in serum or urine. Four of seven metabolites were identified, and were identical with the main metabolites of aminopyrine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Propyphenazon, Pharmaceutical Secondary Standard; Certified Reference Material
Propyphenazon, European Pharmacopoeia (EP) Reference Standard