- Oxymetholone hepatotoxicity enhanced by concomitant use of cyclosporin A in a bone marrow transplant patient.
Oxymetholone hepatotoxicity enhanced by concomitant use of cyclosporin A in a bone marrow transplant patient.
Clinical and laboratory haematology (1994-06-01)
P Wood, J A Yin
PMID7955931
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Stanozolol Unreinheit B, European Pharmacopoeia (EP) Reference Standard