- Drug-induced vanishing bile duct syndrome: response to ursodiol.
Drug-induced vanishing bile duct syndrome: response to ursodiol.
The American journal of gastroenterology (1996-07-01)
C B O'Brien, D S Shields, S H Saul, K R Reddy
PMID8678017
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Ursodesoxycholsäure, European Pharmacopoeia (EP) Reference Standard
Ursodesoxycholsäure für die Systemeignung, European Pharmacopoeia (EP) Reference Standard