Merck
[Fulminant liver failure with a fatal outcome due to flutamide].
Medicina clinica (1997-02-15)
C M Fernández Peña, L E Morano Amado, J Montes Santiago, C Fachal
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Flutamid
Flutamid, European Pharmacopoeia (EP) Reference Standard
Flutamid für die Systemeignung, European Pharmacopoeia (EP) Reference Standard