Merck
  • Lisinopril-induced isolated visceral angioedema: review of ACE-inhibitor-induced small bowel angioedema.
Lisinopril-induced isolated visceral angioedema: review of ACE-inhibitor-induced small bowel angioedema.
Digestive diseases and sciences (1997-04-01)
M F Abdelmalek, D D Douglas
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lisinopril, ≥98% (HPLC)
Supelco
Lisinopril, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Lisinopril, United States Pharmacopeia (USP) Reference Standard
Lisinopril Dihydrat, European Pharmacopoeia (EP) Reference Standard