Merck
[Application for therapeutic use of deuterium oxide (D2O) against human pancreatic cancer].
Gan to kagaku ryoho. Cancer & chemotherapy (1996-10-01)
H Omori, Y Nio, H Takeda, K Tamura
ZUSAMMENFASSUNG
This study was designed to assess the possibility of D2O as a therapeutic agent against human pancreatic cancer. We examined the effect of D2O on growth and invasion of human pancreatic cancer cells (Panc-1, MIA PaCa-2, BxPC-3) by using MTT assay and nude mice, on which human pancreas cancer cells were transplanted. In vitro growth of the three lines was significantly inhibited by D2O in a dose dependent manner, especially at more than a 10% concentration. Growth of Panc-1, which was transplanted in nude mice, was also inhibited by continuous oral intake of 30% D2O. Furthermore, the effect of D2O on cancer invasiveness was assessed by Matrigel invasion chamber assay. BxPC-3, which is the most adherent of the three lines, significantly reduced its invasiveness in more than 30% D2O media. In addition, the present study also demonstrated the inhibition of cancer cell locomotion by D2O in phagocytokinetic analysis. In conclusion, D2O may be applied for the therapeutic use against pancreatic cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuteriumoxid, filtered, 99.8 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.8 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuteriumoxid, 70 atom % D
Sigma-Aldrich
Deuteriumoxid, 60 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D